Assay Details
| Gene Symbol: vps-52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076881.8 | NP_509282.4 | 2 - 3 | 289 | 132 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

vps-52| Gene Symbol: vps-52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076881.8 | NP_509282.4 | 2 - 3 | 289 | 132 | |
Set Membership: |
|