Assay Details
| Gene Symbol: pcyt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038450.4 | NP_001033539.1 | 5 - 6 | 1106 | 108 | |
| NM_001038451.3 | NP_001033540.1 | 4 - 5 | 839 | 108 | ||
More Information
Set Membership: |
|

pcyt-1
Genomic Map