Assay Details
| Gene Symbol: deg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373369.2 | NP_001360711.1 | 13 - 14 | 1556 | 133 | |
| NM_001373370.2 | NP_001360706.1 | 13 | 1814 | 133 | ||
| NM_001373371.2 | NP_001360707.1 | 12 | 1646 | 133 | ||
| NM_001373372.2 | NP_001360708.1 | 11 | 1592 | 133 | ||
| NM_001373373.2 | NP_001360709.1 | 12 | 1784 | 133 | ||
| NM_001373374.2 | NP_001360710.1 | 10 | 1562 | 133 | ||
| NM_076910.5 | NP_509311.1 | 11 - 12 | 1626 | 133 | ||
| GenBank mRNA | L34414.1 | - | 11 - 12 | 1648 | 133 | |
More Information
Set Membership: |
|

deg-1
Genomic Map