Assay Details
| Gene Symbol: deg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373369.2 | NP_001360711.1 | 8 - 9 | 894 | 63 | |
| NM_001373373.2 | NP_001360709.1 | 6 | 954 | 63 | ||
| NM_001373374.2 | NP_001360710.1 | 6 | 954 | 63 | ||
| NM_076910.5 | NP_509311.1 | 6 - 7 | 964 | 63 | ||
| GenBank mRNA | L34414.1 | - | 6 - 7 | 986 | 63 | |
More Information
Set Membership: |
|

deg-1
Genomic Map