Assay Details
| Gene Symbol: deg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373369.2 | NP_001360711.1 | 10 - 11 | 1072 | 169 | |
| NM_001373371.2 | NP_001360707.1 | 9 | 1162 | 169 | ||
| NM_076910.5 | NP_509311.1 | 8 - 9 | 1142 | 169 | ||
| GenBank mRNA | L34414.1 | - | 8 - 9 | 1164 | 169 | |
More Information
Set Membership: |
|

deg-1
Genomic Map