Assay Details
| Gene Symbol: adm-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076917.6 | NP_509318.1 | 5 - 6 | 594 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

adm-4| Gene Symbol: adm-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076917.6 | NP_509318.1 | 5 - 6 | 594 | 109 | |
Set Membership: |
|