Assay Details
| Gene Symbol: dim-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029249.6 | NP_001024420.1 | 5 - 6 | 776 | 104 | |
| GenBank mRNA | AY095447.1 | - | 5 - 6 | 795 | 104 | |
More Information
Set Membership: |
|

dim-1
Genomic Map