Assay Details
| Gene Symbol: hil-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076974.5 | NP_509375.1 | 1 - 2 | 281 | 98 | |
| GenBank mRNA | AF012253.1 | - | 1 - 2 | 260 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hil-3| Gene Symbol: hil-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076974.5 | NP_509375.1 | 1 - 2 | 281 | 98 | |
| GenBank mRNA | AF012253.1 | - | 1 - 2 | 260 | 98 | |
Set Membership: |
|