Assay Details
| Gene Symbol: endu-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076990.7 | NP_509391.1 | 8 - 9 | 1587 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

endu-2| Gene Symbol: endu-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076990.7 | NP_509391.1 | 8 - 9 | 1587 | 78 | |
Set Membership: |
|