Assay Details
| Gene Symbol: sec-16A.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373388.2 | NP_001360732.1 | 13 - 14 | 4509 | 102 | |
| NM_076998.5 | NP_509399.2 | 14 - 15 | 4632 | 102 | ||
More Information
Set Membership: |
|

sec-16A.2
Genomic Map