Assay Details
| Gene Symbol: sec-16A.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029386.5 | NP_001024557.1 | 2 - 3 | 391 | 58 | |
| NM_001373387.3 | NP_001360731.1 | 2 - 3 | 403 | 58 | ||
| NM_001373388.2 | NP_001360732.1 | 2 - 3 | 391 | 58 | ||
| NM_001373389.3 | NP_001360733.1 | 2 - 3 | 391 | 58 | ||
| NM_076998.5 | NP_509399.2 | 2 - 3 | 391 | 58 | ||
| NM_076999.8 | NP_509400.3 | 2 - 3 | 400 | 58 | ||
More Information
Set Membership: |
|

sec-16A.2
Genomic Map