Assay Details
| Gene Symbol: utx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322626.3 | NP_001309685.1 | 4 - 5 | 621 | 130 | |
| NM_001322627.4 | NP_001309686.1 | 4 - 5 | 626 | 130 | ||
More Information
Set Membership: |
|

utx-1
Genomic Map