Assay Details
| Gene Symbol: stg-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029385.5 | NP_001024556.2 | 2 - 3 | 962 | 155 | |
| GenBank mRNA | EU019551.1 | - | 2 - 3 | 317 | 155 | |
More Information
Set Membership: |
|

stg-2
Genomic Map