Assay Details
| Gene Symbol: cyn-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077106.2 | NP_509507.1 | 4 - 5 | 1080 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cyn-8| Gene Symbol: cyn-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077106.2 | NP_509507.1 | 4 - 5 | 1080 | 81 | |
Set Membership: |
|