Assay Details
| Gene Symbol: snf-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077126.3 | NP_509527.1 | 2 - 3 | 720 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

snf-12| Gene Symbol: snf-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077126.3 | NP_509527.1 | 2 - 3 | 720 | 77 | |
Set Membership: |
|