Assay Details
| Gene Symbol: ajm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330906.2 | NP_001317734.1 | 5 - 6 | 653 | 88 | |
| NM_001381045.1 | NP_001368606.1 | 5 - 6 | 616 | 88 | ||
| NM_001381046.2 | NP_001367383.1 | 5 - 6 | 624 | 88 | ||
| NM_001381047.2 | NP_001367382.1 | 5 - 6 | 616 | 88 | ||
| NM_001381048.1 | NP_001367381.1 | 5 - 6 | 653 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001381048.1 transcripts.
Set Membership: |
|

ajm-1
Genomic Map