Assay Details
| Gene Symbol: tmbi-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077142.8 | NP_509543.2 | 3 - 4 | 226 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tmbi-4| Gene Symbol: tmbi-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077142.8 | NP_509543.2 | 3 - 4 | 226 | 65 | |
Set Membership: |
|