Assay Details
| Gene Symbol: oga-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129711.3 | NP_001123183.1 | 11 - 12 | 2126 | 105 | |
| GenBank mRNA | DQ407522.1 | - | 11 - 12 | 2076 | 105 | |
More Information
Set Membership: |
|

oga-1
Genomic Map