Assay Details
| Gene Symbol: fln-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313394.3 | NP_001300323.1 | 5 - 6 | 540 | 80 | |
| NM_001313395.3 | NP_001300324.1 | 5 - 6 | 540 | 80 | ||
| NM_001373422.1 | NP_001359882.1 | 16 - 17 | 2307 | 80 | ||
| NM_001373423.2 | NP_001360663.1 | 16 | 2256 | 80 | ||
| NM_001373424.2 | NP_001360664.1 | 16 | 2256 | 80 | ||
| NM_001373425.2 | NP_001360405.1 | 16 | 2256 | 80 | ||
| NM_001373426.2 | NP_001360665.1 | 16 | 2256 | 80 | ||
| NM_001373427.2 | NP_001360666.1 | 16 | 2256 | 80 | ||
| NM_001373428.2 | NP_001360667.1 | 16 | 2256 | 80 | ||
| NM_001373429.2 | NP_001360668.1 | 16 | 2256 | 80 | ||
| NM_001381052.1 | NP_001368576.1 | 8 - 9 | 1033 | 80 | ||
| NR_132548.1 | 18 - 19 | 2439 | 80 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001381052.1 transcripts.
Set Membership: |
|

fln-2
Genomic Map