Assay Details
| Gene Symbol: fln-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270146.3 | NP_001257075.2 | 20 - 21 | 4984 | 134 | |
| NM_001313395.3 | NP_001300324.1 | 27 - 28 | 5773 | 134 | ||
| NM_001313397.3 | NP_001300326.1 | 22 - 23 | 5248 | 134 | ||
| NM_001313399.3 | NP_001300328.1 | 19 - 20 | 4816 | 134 | ||
| NM_001313401.3 | NP_001300330.1 | 17 - 18 | 4624 | 134 | ||
| NM_001313403.3 | NP_001300332.1 | 12 - 13 | 3901 | 134 | ||
| NM_001373423.2 | NP_001360663.1 | 39 | 7582 | 134 | ||
| NM_001373425.2 | NP_001360405.1 | 38 | 7489 | 134 | ||
| NM_001373427.2 | NP_001360666.1 | 38 | 7498 | 134 | ||
| NM_001373429.2 | NP_001360668.1 | 37 | 7405 | 134 | ||
| NM_001381052.1 | NP_001368576.1 | 31 - 32 | 6359 | 134 | ||
More Information
Set Membership: |
|

fln-2
Genomic Map