Assay Details
| Gene Symbol: fln-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270145.4 | NP_001257074.2 | 3 - 4 | 353 | 64 | |
| NM_001270146.3 | NP_001257075.2 | 3 - 4 | 353 | 64 | ||
| NM_001313394.3 | NP_001300323.1 | 10 - 11 | 1142 | 64 | ||
| NM_001313395.3 | NP_001300324.1 | 10 - 11 | 1142 | 64 | ||
| NM_001313396.3 | NP_001300325.1 | 5 - 6 | 617 | 64 | ||
| NM_001313397.3 | NP_001300326.1 | 5 - 6 | 617 | 64 | ||
| NM_001313398.3 | NP_001300327.1 | 2 - 3 | 185 | 64 | ||
| NM_001313399.3 | NP_001300328.1 | 2 - 3 | 185 | 64 | ||
| NM_001373422.1 | NP_001359882.1 | 21 - 22 | 2909 | 64 | ||
| NM_001373423.2 | NP_001360663.1 | 21 | 2858 | 64 | ||
| NM_001373424.2 | NP_001360664.1 | 21 | 2858 | 64 | ||
| NM_001373425.2 | NP_001360405.1 | 21 | 2858 | 64 | ||
| NM_001373426.2 | NP_001360665.1 | 20 | 2774 | 64 | ||
| NM_001373427.2 | NP_001360666.1 | 20 | 2774 | 64 | ||
| NM_001373428.2 | NP_001360667.1 | 20 | 2774 | 64 | ||
| NM_001373429.2 | NP_001360668.1 | 20 | 2774 | 64 | ||
| NM_001381052.1 | NP_001368576.1 | 13 - 14 | 1635 | 64 | ||
| NR_132548.1 | 22 - 23 | 3362 | 64 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001381052.1 transcripts.
Set Membership: |
|

fln-2
Genomic Map