Assay Details
| Gene Symbol: fln-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270145.4 | NP_001257074.2 | 8 - 9 | 969 | 67 | |
| NM_001270146.3 | NP_001257075.2 | 8 - 9 | 969 | 67 | ||
| NM_001313394.3 | NP_001300323.1 | 15 - 16 | 1758 | 67 | ||
| NM_001313395.3 | NP_001300324.1 | 15 - 16 | 1758 | 67 | ||
| NM_001313396.3 | NP_001300325.1 | 10 - 11 | 1233 | 67 | ||
| NM_001313397.3 | NP_001300326.1 | 10 - 11 | 1233 | 67 | ||
| NM_001313398.3 | NP_001300327.1 | 7 - 8 | 801 | 67 | ||
| NM_001313399.3 | NP_001300328.1 | 7 - 8 | 801 | 67 | ||
| NM_001313400.3 | NP_001300329.1 | 5 - 6 | 609 | 67 | ||
| NM_001313401.3 | NP_001300330.1 | 5 - 6 | 609 | 67 | ||
| NM_001373422.1 | NP_001359882.1 | 26 - 27 | 3525 | 67 | ||
| NM_001373423.2 | NP_001360663.1 | 26 | 3474 | 67 | ||
| NM_001373424.2 | NP_001360664.1 | 26 | 3474 | 67 | ||
| NM_001373425.2 | NP_001360405.1 | 26 | 3474 | 67 | ||
| NM_001373426.2 | NP_001360665.1 | 25 | 3390 | 67 | ||
| NM_001373427.2 | NP_001360666.1 | 25 | 3390 | 67 | ||
| NM_001373428.2 | NP_001360667.1 | 25 | 3390 | 67 | ||
| NM_001373429.2 | NP_001360668.1 | 25 | 3390 | 67 | ||
| NM_001381052.1 | NP_001368576.1 | 18 - 19 | 2251 | 67 | ||
| NR_132548.1 | 27 - 28 | 3978 | 67 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001381052.1 transcripts.
Set Membership: |
|

fln-2
Genomic Map