Assay Details
| Gene Symbol: tba-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077190.5 | NP_509591.2 | 7 - 8 | 1076 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tba-8| Gene Symbol: tba-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077190.5 | NP_509591.2 | 7 - 8 | 1076 | 62 | |
Set Membership: |
|