Assay Details
| Gene Symbol: him-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029411.3 | NP_001024582.1 | 36 - 37 | 9157 | 105 | |
| NM_001373449.3 | NP_001359724.1 | 36 | 9096 | 105 | ||
| NM_001373450.2 | NP_001360633.1 | 35 - 36 | 8802 | 105 | ||
| NM_001373451.2 | NP_001360634.1 | 35 - 36 | 8802 | 105 | ||
| NM_077234.7 | NP_509635.2 | 36 - 37 | 9096 | 105 | ||
| GenBank mRNA | AF074901.1 | - | 36 - 37 | 9096 | 105 | |
More Information
Set Membership: |
|

him-4
Genomic Map