Assay Details
| Gene Symbol: him-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029411.3 | NP_001024582.1 | 61 - 62 | 15458 | 107 | |
| NM_001129660.3 | NP_001123132.1 | 3 - 4 | 439 | 107 | ||
| NM_001373450.2 | NP_001360633.1 | 60 - 61 | 15103 | 107 | ||
| NM_001373451.2 | NP_001360634.1 | 60 - 61 | 15034 | 107 | ||
| NM_077234.7 | NP_509635.2 | 61 - 62 | 15328 | 107 | ||
| GenBank mRNA | AF074901.1 | - | 61 - 62 | 15397 | 107 | |
More Information
Set Membership: |
|

him-4
Genomic Map