Assay Details
| Gene Symbol: dpy-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077244.4 | NP_509645.1 | 10 - 11 | 4599 | 129 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dpy-22| Gene Symbol: dpy-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077244.4 | NP_509645.1 | 10 - 11 | 4599 | 129 | |
Set Membership: |
|