Assay Details
| Gene Symbol: rrc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029512.5 | NP_001024683.1 | 12 - 13 | 2145 | 75 | |
| NM_001029513.6 | NP_001024684.1 | 12 - 13 | 2165 | 75 | ||
| NM_001270161.4 | NP_001257090.1 | 12 - 13 | 2138 | 75 | ||
More Information
Set Membership: |
|

rrc-1
Genomic Map