Assay Details
| Gene Symbol: rrc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029512.5 | NP_001024683.1 | 10 - 11 | 1728 | 68 | |
| NM_001029513.6 | NP_001024684.1 | 10 - 11 | 1748 | 68 | ||
More Information
Set Membership: |
|

rrc-1
Genomic Map