Assay Details
| Gene Symbol: glo-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029666.3 | NP_001024837.1 | 3 - 4 | 396 | 76 | |
| GenBank mRNA | DQ016970.1 | - | 3 - 4 | 390 | 76 | |
More Information
Set Membership: |
|

glo-1
Genomic Map