Assay Details
| Gene Symbol: mgl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129721.4 | NP_001123193.2 | 17 - 18 | 2511 | 102 | |
| NM_001313458.3 | NP_001300387.1 | 18 - 19 | 2553 | 102 | ||
| NM_001313459.3 | NP_001300388.1 | 14 - 15 | 2286 | 102 | ||
| NM_077273.4 | NP_509674.2 | 18 - 19 | 2770 | 102 | ||
More Information
Set Membership: |
|

mgl-1
Genomic Map