Assay Details
| Gene Symbol: mgl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129721.4 | NP_001123193.2 | 6 - 7 | 581 | 107 | |
| NM_001313458.3 | NP_001300387.1 | 7 - 8 | 623 | 107 | ||
| NM_001313459.3 | NP_001300388.1 | 3 - 4 | 356 | 107 | ||
| NM_077273.4 | NP_509674.2 | 7 - 8 | 840 | 107 | ||
More Information
Set Membership: |
|

mgl-1
Genomic Map