Assay Details
| Gene Symbol: cbs-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270162.3 | NP_001257091.1 | 1 - 2 | 56 | 79 | |
| NM_001270163.4 | NP_001257092.1 | 1 - 2 | 62 | 79 | ||
More Information
Set Membership: |
|

cbs-1
Genomic Map