Assay Details
| Gene Symbol: mlck-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077288.6 | NP_509689.1 | 14 - 15 | 2483 | 150 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mlck-1| Gene Symbol: mlck-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077288.6 | NP_509689.1 | 14 - 15 | 2483 | 150 | |
Set Membership: |
|