Assay Details
| Gene Symbol: pkn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270173.3 | NP_001257102.1 | 1 - 2 | 280 | 123 | |
| NM_001373472.1 | NP_001360641.1 | 1 - 2 | 274 | 123 | ||
More Information
Set Membership: |
|

pkn-1
Genomic Map