Assay Details
| Gene Symbol: syd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077367.9 | NP_509768.1 | 12 - 13 | 1801 | 103 | |
| GenBank mRNA | AF170122.1 | - | 11 - 12 | 1728 | 103 | |
More Information
Set Membership: |
|

syd-2
Genomic Map