Assay Details
| Gene Symbol: syd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077367.9 | NP_509768.1 | 20 - 21 | 3436 | 141 | |
| GenBank mRNA | AF170122.1 | - | 19 - 20 | 3363 | 141 | |
More Information
Set Membership: |
|

syd-2
Genomic Map