Assay Details
| Gene Symbol: syd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077367.9 | NP_509768.1 | 3 - 4 | 529 | 66 | |
| GenBank mRNA | AF170122.1 | - | 2 - 3 | 456 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

syd-2| Gene Symbol: syd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077367.9 | NP_509768.1 | 3 - 4 | 529 | 66 | |
| GenBank mRNA | AF170122.1 | - | 2 - 3 | 456 | 66 | |
Set Membership: |
|