Assay Details
| Gene Symbol: syd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077367.9 | NP_509768.1 | 7 - 8 | 980 | 71 | |
| GenBank mRNA | AF170122.1 | - | 6 - 7 | 907 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

syd-2| Gene Symbol: syd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077367.9 | NP_509768.1 | 7 - 8 | 980 | 71 | |
| GenBank mRNA | AF170122.1 | - | 6 - 7 | 907 | 71 | |
Set Membership: |
|