Assay Details
| Gene Symbol: syd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077367.9 | NP_509768.1 | 10 - 11 | 1404 | 68 | |
| GenBank mRNA | AF170122.1 | - | 9 - 10 | 1331 | 68 | |
More Information
Set Membership: |
|

syd-2
Genomic Map