Assay Details
| Gene Symbol: plpp-1.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077389.4 | NP_509790.2 | 3 - 4 | 263 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

plpp-1.3| Gene Symbol: plpp-1.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077389.4 | NP_509790.2 | 3 - 4 | 263 | 76 | |
Set Membership: |
|