Assay Details
| Gene Symbol: plpp-1.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077389.4 | NP_509790.2 | 6 - 7 | 540 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

plpp-1.3| Gene Symbol: plpp-1.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077389.4 | NP_509790.2 | 6 - 7 | 540 | 114 | |
Set Membership: |
|