Assay Details
| Gene Symbol: mbr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077393.7 | NP_509794.2 | 9 - 10 | 1243 | 77 | |
| GenBank mRNA | AB236333.1 | - | 8 - 9 | 1147 | 77 | |
More Information
Set Membership: |
|

mbr-1
Genomic Map