Assay Details
Gene Symbol: plc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029446.3 | NP_001024617.1 | 10 - 11 | 1813 | 66 | |
NM_001029448.6 | NP_001024619.1 | 10 - 11 | 1775 | 66 | ||
NM_001129669.4 | NP_001123141.2 | 43 - 44 | 6653 | 66 | ||
NM_001136454.3 | NP_001129926.1 | 10 - 11 | 1805 | 66 | ||
NM_001136455.4 | NP_001129927.1 | 10 - 11 | 1997 | 66 | ||
NM_001373489.2 | NP_001360637.1 | 43 - 44 | 6632 | 66 | ||
NM_001373491.2 | NP_001360639.1 | 43 - 44 | 6662 | 66 | ||
NM_001373492.2 | NP_001360635.1 | 43 - 44 | 6683 | 66 | ||
NM_001373493.2 | NP_001360636.1 | 44 - 45 | 6803 | 66 | ||
NM_001373494.2 | NP_001359720.1 | 44 - 45 | 6824 | 66 | ||
NM_001373495.2 | NP_001360638.1 | 44 - 45 | 6833 | 66 | ||
NM_077404.6 | NP_509805.3 | 10 - 11 | 1775 | 66 | ||
GenBank mRNA | AF044576.1 | - | 10 - 11 | 1835 | 66 | |
AM992255.1 | - | 10 - 11 | 1805 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
