Assay Details
| Gene Symbol: plc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029446.3 | NP_001024617.1 | 16 - 17 | 2868 | 80 | |
| NM_001029448.6 | NP_001024619.1 | 16 - 17 | 2830 | 80 | ||
| NM_001129669.4 | NP_001123141.2 | 49 - 50 | 7708 | 80 | ||
| NM_001136454.3 | NP_001129926.1 | 16 - 17 | 2860 | 80 | ||
| NM_001136455.4 | NP_001129927.1 | 16 - 17 | 3052 | 80 | ||
| NM_001373489.2 | NP_001360637.1 | 49 - 50 | 7687 | 80 | ||
| NM_001373491.2 | NP_001360639.1 | 49 - 50 | 7717 | 80 | ||
| NM_001373492.2 | NP_001360635.1 | 49 - 50 | 7738 | 80 | ||
| NM_001373493.2 | NP_001360636.1 | 50 - 51 | 7858 | 80 | ||
| NM_001373494.2 | NP_001359720.1 | 50 - 51 | 7879 | 80 | ||
| NM_001373495.2 | NP_001360638.1 | 50 - 51 | 7888 | 80 | ||
| NM_077404.6 | NP_509805.3 | 16 - 17 | 2830 | 80 | ||
| GenBank mRNA | AF044576.1 | - | 16 - 17 | 2890 | 80 | |
| AM992255.1 | - | 16 - 17 | 2860 | 80 | ||
More Information
Set Membership: |
|

plc-1
Genomic Map