Assay Details
| Gene Symbol: plc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029446.3 | NP_001024617.1 | 27 - 28 | 5033 | 94 | |
| NM_001029448.6 | NP_001024619.1 | 27 - 28 | 4995 | 94 | ||
| NM_001129669.4 | NP_001123141.2 | 60 - 61 | 9873 | 94 | ||
| NM_001136454.3 | NP_001129926.1 | 27 - 28 | 5025 | 94 | ||
| NM_001136455.4 | NP_001129927.1 | 27 - 28 | 5217 | 94 | ||
| NM_001373489.2 | NP_001360637.1 | 60 - 61 | 9852 | 94 | ||
| NM_001373491.2 | NP_001360639.1 | 60 - 61 | 9882 | 94 | ||
| NM_001373492.2 | NP_001360635.1 | 60 - 61 | 9903 | 94 | ||
| NM_001373493.2 | NP_001360636.1 | 61 - 62 | 10023 | 94 | ||
| NM_001373494.2 | NP_001359720.1 | 61 - 62 | 10044 | 94 | ||
| NM_001373495.2 | NP_001360638.1 | 61 - 62 | 10053 | 94 | ||
| NM_077404.6 | NP_509805.3 | 27 - 28 | 4995 | 94 | ||
| GenBank mRNA | AF044576.1 | - | 27 - 28 | 5055 | 94 | |
| AM992255.1 | - | 27 - 28 | 5025 | 94 | ||
More Information
Set Membership: |
|

plc-1
Genomic Map