Assay Details
| Gene Symbol: plc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129669.4 | NP_001123141.2 | 20 - 21 | 2412 | 81 | |
| NM_001373489.2 | NP_001360637.1 | 20 - 21 | 2412 | 81 | ||
| NM_001373491.2 | NP_001360639.1 | 20 - 21 | 2412 | 81 | ||
| NM_001373492.2 | NP_001360635.1 | 20 - 21 | 2412 | 81 | ||
| NM_001373493.2 | NP_001360636.1 | 21 - 22 | 2583 | 81 | ||
| NM_001373494.2 | NP_001359720.1 | 21 - 22 | 2583 | 81 | ||
| NM_001373495.2 | NP_001360638.1 | 21 - 22 | 2583 | 81 | ||
More Information
Set Membership: |
|

plc-1
Genomic Map