Assay Details
| Gene Symbol: pgp-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270187.3 | NP_001257116.1 | 12 - 13 | 2203 | 122 | |
| NM_001270188.3 | NP_001257117.1 | 9 - 10 | 1860 | 122 | ||
More Information
Set Membership: |
|

pgp-5
Genomic Map