Assay Details
| Gene Symbol: pgp-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270187.3 | NP_001257116.1 | 16 - 17 | 3260 | 113 | |
| NM_001270188.3 | NP_001257117.1 | 13 - 14 | 2917 | 113 | ||
More Information
Set Membership: |
|

pgp-5
Genomic Map