Assay Details
| Gene Symbol: pgp-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270187.3 | NP_001257116.1 | 4 - 5 | 409 | 80 | |
| NM_001270188.3 | NP_001257117.1 | 1 - 2 | 66 | 80 | ||
More Information
Set Membership: |
|

pgp-5
Genomic Map