Assay Details
| Gene Symbol: pgp-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270187.3 | NP_001257116.1 | 5 - 6 | 547 | 141 | |
| NM_001270188.3 | NP_001257117.1 | 2 - 3 | 204 | 141 | ||
More Information
Set Membership: |
|

pgp-5
Genomic Map